missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-Dihydroxy-6-methylnicotinate 98.0+%, TCI America™

Catalog No. p-7129193
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08691G 1 g
E08695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08691G Supplier TCI America Supplier No. E08691G

Chemical Identifiers

CAS 70254-52-3
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD02929397
InChI Key CMCZAWPDHHYFPU-UHFFFAOYSA-N
PubChem CID 54676534
IUPAC Name ethyl 4-hydroxy-6-methyl-2-oxo-1H-pyridine-3-carboxylate
SMILES CCOC(=O)C1=C(C=C(NC1=O)C)O

Specifications

Melting Point 210°C
Color White
Quantity 1 g
Synonym ethyl 2,4-dihydroxy-6-methylnicotinate, ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate, ethyl 2,4-dihydroxy-6-methylpyridine-3-carboxylate, 2,4-dihydroxy-6-methylnicotinic acid ethyl ester, ethyl2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, ethyl 2,4-dihydroxy-6-methyl-3-pyridine-carboxylate, 2,4-dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, 4-hydroxy-6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid ethyl ester, 2,4-dihydroxy-6-methyl-nicotinic acid ethyl ester
Formula Weight 197.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2,4-Dihydroxy-6-methylnicotinate