accessibility menu, dialog, popup

UserName

H3COOHNOCH3OH

CAS RN 70254-52-3

IUPAC Name: ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Synonyms: ethyl 4-hydroxy-6-methyl-2-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 197.19
Molecular Formula: C9H11NO4
InChi Key: CMCZAWPDHHYFPU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(O)C=C(C)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results