missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-Dichloropyrimidine-5-carboxylate 98.0+%, TCI America™

Catalog No. E11541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11541G 1 g
E11545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11541G Supplier TCI America Supplier No. E11541G
Only null left

Chemical Identifiers

CAS 51940-64-8
Molecular Formula C7H6Cl2N2O2
Molecular Weight (g/mol) 221.037
MDL Number MFCD09910281
InChI Key SRJBDGLSCPDXBL-UHFFFAOYSA-N
Synonym ethyl 2,4-dichloro-5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester, ethyl 2,4-dichloro-pyrimidine-5-carboxylate, 2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonylpyrimidine, 2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloropyrimidine-5-carboxylic acid ethyl ester, pubchem24010, ethyl2,4-dichloro-5-pyrimidinecarboxylate
PubChem CID 104020
IUPAC Name ethyl 2,4-dichloropyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)Cl

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 145°C
Quantity 1 g
Formula Weight 221.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2,4-Dichloropyrimidine-5-carboxylate
TSCA No
Recommended Storage Refrigerator