accessibility menu, dialog, popup

UserName

CAS RN 51940-64-8

IUPAC Name: ethyl 2,4-dichloropyrimidine-5-carboxylate
Synonyms: ethyl 2,4-dichloropyrimidine-5-carboxylate
Molecular Weight (g/mol): 221.04
Molecular Formula: C7H6Cl2N2O2
InChi Key: SRJBDGLSCPDXBL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(Cl)N=C1Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)