missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%

Catalog No. p-7056046
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6478803 1 g
AAH6478806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6478803 Supplier Thermo Scientific Chemicals Supplier No. H6478803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51940-64-8 | C7H6Cl2N2O2 | 221.037 g/mol

It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 51940-64-8
Molecular Formula C7H6Cl2N2O2
Molecular Weight (g/mol) 221.037
MDL Number MFCD09910281
InChI Key SRJBDGLSCPDXBL-UHFFFAOYSA-N
PubChem CID 104020
IUPAC Name ethyl 2,4-dichloropyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)Cl

Specifications

Melting Point 36°C to 37°C
Color White
Quantity 1 g
Beilstein 152173
Synonym ethyl 2,4-dichloro-5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester, ethyl 2,4-dichloro-pyrimidine-5-carboxylate, 2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonylpyrimidine, 2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloropyrimidine-5-carboxylic acid ethyl ester, pubchem24010, ethyl2,4-dichloro-5-pyrimidinecarboxylate
Solubility Information Slightly soluble in water.
Formula Weight 221.04
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Ethyl 2,4-dichloropyrimidine-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 257-531-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only