missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,3-Butadienoate 95.0+%, TCI America™
$171.49 - $685.97
Chemical Identifiers
| CAS | 14369-81-4 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.128 |
| MDL Number | MFCD01863567 |
| InChI Key | GLSUOACRAMLJIW-UHFFFAOYSA-N |
| Synonym | 2,3-Butadienoic Acid Ethyl Ester |
| PubChem CID | 4066088 |
| IUPAC Name | ethyl buta-2,3-dienoate |
| SMILES | CCOC(=O)C=C=C |
Chemical Identifiers
| 14369-81-4 | |
| 112.128 | |
| GLSUOACRAMLJIW-UHFFFAOYSA-N | |
| 4066088 | |
| CCOC(=O)C=C=C |
| C6H8O2 | |
| MFCD01863567 | |
| 2,3-Butadienoic Acid Ethyl Ester | |
| ethyl buta-2,3-dienoate |
Specifications
| 14369-81-4 | |
| 80°C | |
| MFCD01863567 | |
| 3272 | |
| GLSUOACRAMLJIW-UHFFFAOYSA-N | |
| ethyl buta-2,3-dienoate | |
| 4066088 | |
| ≥95.0% (GC) | |
| Ethyl 2,3-Butadienoate |
| Yellow | |
| C6H8O2 | |
| 1 g | |
| 2,3-Butadienoic Acid Ethyl Ester | |
| CCOC(=O)C=C=C | |
| 112.128 | |
| 112.13 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Freezer