CAS RN 14369-81-4
IUPAC Name:
ethyl buta-2,3-dienoate
Synonyms:
ethyl buta-2,3-dienoate
Molecular Weight (g/mol):
112.13
Molecular Formula:
C6H8O2
InChi Key:
GLSUOACRAMLJIW-UHFFFAOYNA-N
SMILES:
CCOC(=O)C=C=C
1
–
1
of
1
results
1
Ethyl 2,3-Butadienoate 95.0+%, TCI America™
CAS: 14369-81-4 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.128 MDL Number: MFCD01863567 InChI Key: GLSUOACRAMLJIW-UHFFFAOYSA-N Synonym: 2,3-Butadienoic Acid Ethyl Ester PubChem CID: 4066088 IUPAC Name: ethyl buta-2,3-dienoate SMILES: CCOC(=O)C=C=C