missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,3-Butadienoate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 80°C |
| Quantity | 1 g |
| UN Number | 3272 |
| Formula Weight | 112.13 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl 2,3-Butadienoate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |