missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(2-Methyl-1,3-dioxolan-2-yl)acetate 98.0+%, TCI America™

Catalog No. E0929500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0929500G 500 g
E092925G 25 g
E0929100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0929500G Supplier TCI America Supplier No. E0929500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6413-10-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00152488
InChI Key XWEOGMYZFCHQNT-UHFFFAOYSA-N
Synonym 2-(2-Methyl-1,3-dioxolan-2-yl)acetic Acid Ethyl Ester
PubChem CID 80865
IUPAC Name ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate
SMILES CCOC(=O)CC1(OCCO1)C

Specifications

Color Colorless
Boiling Point 98°C
Quantity 500 g
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-(2-Methyl-1,3-dioxolan-2-yl)acetate
EINECSNumber (5)-1178
RTECSNumber JH6762500
TSCA Yes