accessibility menu, dialog, popup

UserName

CAS RN 6413-10-1

IUPAC Name: ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate
Synonyms: fructone ethylacetoacetate ethylene glycol ketal
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: XWEOGMYZFCHQNT-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1(C)OCCO1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results