missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate 95.0+%, TCI America™
$106.33 - $320.87
Chemical Identifiers
| CAS | 97-41-6 |
|---|---|
| Molecular Formula | C12H20O2 |
| Molecular Weight (g/mol) | 196.29 |
| MDL Number | MFCD00001304 |
| InChI Key | VIMXTGUGWLAOFZ-UHFFFAOYSA-N |
| Synonym | ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate |
| PubChem CID | 7334 |
| IUPAC Name | ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate |
| SMILES | CCOC(=O)C1C(C1(C)C)C=C(C)C |
Chemical Identifiers
| C12H20O2 | |
| MFCD00001304 | |
| ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate | |
| ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate |
Specifications
| 97-41-6 | |
| 112°C | |
| MFCD00001304 | |
| VIMXTGUGWLAOFZ-UHFFFAOYSA-N | |
| ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate | |
| 7334 | |
| ≥95.0% (GC) | |
| Ethyl 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate |
| Yellow | |
| C12H20O2 | |
| ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate | |
| CCOC(=O)C1C(C1(C)C)C=C(C)C | |
| 196.29 | |
| 196.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2443
RTECSNumber : GZ1727000
TSCA : Yes