missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate 97.0+%, TCI America™

Catalog No. E13141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13141G 1 g
E13145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E13141G Supplier TCI America Supplier No. E13141G

Chemical Identifiers

CAS 205865-67-4
Molecular Formula C7H14F2O2Si
Molecular Weight (g/mol) 196.269
MDL Number MFCD04973092
InChI Key DYAKYYSMROBYNG-UHFFFAOYSA-N
Synonym 2,2-Difluoro-2-(trimethylsilyl)acetic Acid Ethyl Ester
PubChem CID 2758940
IUPAC Name ethyl 2,2-difluoro-2-trimethylsilylacetate
SMILES CCOC(=O)C(F)(F)[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 55°C
Quantity 1 g
UN Number 3272
Formula Weight 196.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate