missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,2-diethylacetoacetate, 97%

Catalog No. AAB2372906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2372906 5 g
AAB2372914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2372906 Supplier Thermo Scientific Chemicals Supplier No. B2372906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1619-57-4 | C10H18O3 | 186.251 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1619-57-4
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD00048552
InChI Key WEIQRLLXVVSKIL-UHFFFAOYSA-N
Synonym ethyl 2,2-diethylacetoacetate, butanoic acid, 2,2-diethyl-3-oxo-, ethyl ester, diethyl acetoacetate, acmc-20al1q, acetoacetic acid, 2,2-diethyl-, ethyl ester, 2,2-diethylacetoacetic acid ethyl ester, 2-ethyl-2-acetylbutyric acid ethyl ester, 2,2-diethyl-3-oxo-butyric acid ethyl ester, butanoic acid,2,2-diethyl-3-oxo-, ethyl ester
PubChem CID 538781
IUPAC Name ethyl 2,2-diethyl-3-oxobutanoate
SMILES CCC(CC)(C(=O)C)C(=O)OCC

Specifications

Boiling Point 84°C to 86°C (13 mmHg)
Refractive Index 1.433
Quantity 5 g
Beilstein 1767891
Formula Weight 186.25
Percent Purity 97%
Chemical Name or Material Ethyl 2,2-diethylacetoacetate
EINECSNumber 216-581-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only