missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1H-Tetrazole-5-acetate 98.0+%, TCI America™
Supplier: TCI America E085925G
Specifications
| Ethyl 1H-Tetrazole-5-acetate | |
| 128°C | |
| C5H8N4O2 | |
| 25 g | |
| NAOHMNNTUFFTBF-UHFFFAOYSA-N | |
| ethyl 2-(2H-tetrazol-5-yl)acetate | |
| 543616 | |
| ≥98.0% (HPLC,T) |
| 13616-37-0 | |
| White | |
| MFCD00274213 | |
| 1H-Tetrazole-5-acetic Acid Ethyl Ester | |
| CCOC(=O)CC1=NNN=N1 | |
| 156.145 | |
| 156.15 | |
| Crystalline Powder |
Chemical Identifiers
| 13616-37-0 | |
| 156.145 | |
| NAOHMNNTUFFTBF-UHFFFAOYSA-N | |
| 543616 | |
| CCOC(=O)CC1=NNN=N1 |
| C5H8N4O2 | |
| MFCD00274213 | |
| 1H-Tetrazole-5-acetic Acid Ethyl Ester | |
| ethyl 2-(2H-tetrazol-5-yl)acetate |
Safety and Handling
TSCA : No