CAS RN 13616-37-0
CAS RN 13616-37-0
IUPAC Name:
ethyl 2-(2H-1,2,3,4-tetrazol-5-yl)acetate
Synonyms:
ethyl 2-(2H-1,2,3,4-tetrazol-5-yl)acetate
Molecular Weight (g/mol):
156.15
Molecular Formula:
C5H8N4O2
InChi Key:
NAOHMNNTUFFTBF-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=NNN=N1
1
–
1
of
1
results
1
Ethyl 1H-Tetrazole-5-acetate 98.0+%, TCI America™
CAS: 13616-37-0 Molecular Formula: C5H8N4O2 Molecular Weight (g/mol): 156.145 MDL Number: MFCD00274213 InChI Key: NAOHMNNTUFFTBF-UHFFFAOYSA-N Synonym: 1H-Tetrazole-5-acetic Acid Ethyl Ester PubChem CID: 543616 IUPAC Name: ethyl 2-(2H-tetrazol-5-yl)acetate SMILES: CCOC(=O)CC1=NNN=N1