missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate 98.0+%, TCI America™
$61.61 - $177.11
Chemical Identifiers
| CAS | 130250-54-3 |
|---|---|
| Molecular Formula | C13H23NO4 |
| Molecular Weight (g/mol) | 257.33 |
| MDL Number | MFCD04116274 |
| InChI Key | YCXCRFGBFZTUSU-UHFFFAOYSA-N |
| Synonym | ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester |
| PubChem CID | 357727 |
| IUPAC Name | 1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate |
| SMILES | CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C |
Chemical Identifiers
| 130250-54-3 | |
| 257.33 | |
| YCXCRFGBFZTUSU-UHFFFAOYSA-N | |
| 357727 | |
| CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C |
| C13H23NO4 | |
| MFCD04116274 | |
| ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester | |
| 1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate |
Specifications
| 130250-54-3 | |
| Yellow | |
| C13H23NO4 | |
| 5 g | |
| YCXCRFGBFZTUSU-UHFFFAOYSA-N | |
| 1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate | |
| 357727 | |
| ≥98.0% (GC) | |
| Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate |
| 37°C | |
| 93°C | |
| MFCD04116274 | |
| ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester | |
| CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C | |
| 257.33 | |
| 257.33 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No