missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate 98.0+%, TCI America™

Supplier:  TCI America E095425G

 View more versions of this product

Catalog No. E095425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate
37°C
93°C
MFCD04116274
ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester
CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C
257.33
257.33
Crystalline Lumps
130250-54-3
Yellow
C13H23NO4
25 g
YCXCRFGBFZTUSU-UHFFFAOYSA-N
1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate
357727
≥98.0% (GC)

Chemical Identifiers

130250-54-3
257.33
YCXCRFGBFZTUSU-UHFFFAOYSA-N
357727
CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C
C13H23NO4
MFCD04116274
ethyl 1-boc-3-piperidinecarboxylate, 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl n-boc-piperidine-3-carboxylate, ethyl n-boc-3-piperidinecarboxylate, ethyl1-boc-3-piperidinecarboxylate, 1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl-n-boc-piperidine-3-carboxylate, 1-tert-butyl 3-ethyl tetrahydro-1,3 2h-pyridinedicarboxylate, 1,3-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 3-ethyl ester, piperidine-1,3-dicarboxylic acid 1-tert-butyl 3-ethyl ester
1-O-tert-butyl 3-O-ethyl piperidine-1,3-dicarboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions