missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-naphthoate, 97%

Catalog No. AAL0776018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0776018 50 g
AAL0776009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0776018 Supplier Thermo Scientific Chemicals Supplier No. L0776018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3007-97-4 | C13H12O2 | 200.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3007-97-4
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00016805
InChI Key XCTLDQQOHIEUCJ-UHFFFAOYSA-N
Synonym ethyl 1-naphthoate, ethyl alpha-naphthoate, 1-naphthoic acid, ethyl ester, ethyl alfa-naphthoate, 1-naphthalenecarboxylic acid, ethyl ester, 1-naphthoic acid ethyl ester, naphthalene-1-carboxylic acid ethyl ester, ethyl naphthalenecarboxylate, ethyl a-naphthoate, ethylalpha-naphthoate
PubChem CID 76364
IUPAC Name ethyl naphthalene-1-carboxylate
SMILES CCOC(=O)C1=CC=CC2=CC=CC=C21

Specifications

Density 1.11
Boiling Point 308°C to 310°C
Refractive Index 1.593
Quantity 50 g
Beilstein 2047857
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material Ethyl 1-naphthoate
EINECSNumber 221-120-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only