accessibility menu, dialog, popup

UserName

H3COO

CAS RN 3007-97-4

IUPAC Name: ethyl naphthalene-1-carboxylate
Synonyms: ethyl naphthalene-1-carboxylate
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: XCTLDQQOHIEUCJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results