missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Naphthaleneacetate 96.0+%, TCI America™

Supplier:  TCI America N055425G

Catalog No. N055425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 1-Naphthaleneacetate
Yellow
C14H14O2
25 g
XIDPSKQLXKCVQN-UHFFFAOYSA-N
ethyl 2-naphthalen-1-ylacetate
62428
≥96.0% (GC)
2122-70-5
231°C
MFCD00021579
1-Naphthaleneacetic Acid Ethyl Ester
CCOC(=O)CC1=CC=CC2=CC=CC=C21
214.264
214.26
Liquid

Chemical Identifiers

2122-70-5
214.264
XIDPSKQLXKCVQN-UHFFFAOYSA-N
62428
CCOC(=O)CC1=CC=CC2=CC=CC=C21
C14H14O2
MFCD00021579
1-Naphthaleneacetic Acid Ethyl Ester
ethyl 2-naphthalen-1-ylacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : QJ0948000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions