accessibility menu, dialog, popup

UserName

CAS RN 2122-70-5

H3COO

CAS RN 2122-70-5

IUPAC Name: ethyl 2-(naphthalen-1-yl)acetate
Synonyms: ethyl 2-(naphthalen-1-yl)acetate
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: XIDPSKQLXKCVQN-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results