missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-methylimidazole-2-carboxylate, 95%, Thermo Scientific™
$1025.64 - $1025.64
Chemical Identifiers
| CAS | 30148-21-1 |
|---|---|
| Molecular Formula | C7H10N2O2 |
| Molecular Weight (g/mol) | 154.17 |
| MDL Number | MFCD02683326 |
| InChI Key | NOTZYDYZBOBDFE-UHFFFAOYSA-N |
| Synonym | ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester |
| PubChem CID | 4261883 |
| IUPAC Name | ethyl 1-methylimidazole-2-carboxylate |
| SMILES | CCOC(=O)C1=NC=CN1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6118514
|
Thermo Scientific Chemicals
H6118514 |
25 g |
Each for $1,025.64
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEthyl 1-methylimidazole-2-carboxylate is used as a synthetic intermediate for solid phase synthesis of polyamides containing imidazole.
Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 30148-21-1 | |
| 154.17 | |
| NOTZYDYZBOBDFE-UHFFFAOYSA-N | |
| 4261883 | |
| CCOC(=O)C1=NC=CN1C |
| C7H10N2O2 | |
| MFCD02683326 | |
| ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester | |
| ethyl 1-methylimidazole-2-carboxylate |
Specifications
| 30148-21-1 | |
| MFCD02683326 | |
| ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester | |
| NOTZYDYZBOBDFE-UHFFFAOYSA-N | |
| ethyl 1-methylimidazole-2-carboxylate | |
| 4261883 | |
| 95% | |
| Ethyl 1-methylimidazole-2-carboxylate |
| C7H10N2O2 | |
| 25 g | |
| Soluble in dichloromethane,ether,ethyl acetate and methanol. | |
| CCOC(=O)C1=NC=CN1C | |
| 154.17 | |
| 154.17 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures