missing translation for 'onlineSavingsMsg'
Learn More

Éthyle 1-Hydroxy-1H-1,2,3-triazole-4-carboxylate 98,0+%, TCI America™

Numéro de catalogue. p-7129213
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E09011G 1 g
E09015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E09011G Fournisseur TCI America Code fournisseur E09011G

Identifiants chimiques

CAS 137156-41-3
Molecular Formula C5H7N3O3
Molecular Weight (g/mol) 157.129
MDL Number MFCD06796746
InChI Key FIRHQRGFVOSDDY-UHFFFAOYSA-N
Synonym 1-Hydroxy-1H-1,2,3-triazole-4-carboxylic Acid Ethyl Ester
PubChem CID 5270538
IUPAC Name Éthyle 1-hydroxytriazole-4-carboxylate
SMILES CCOC(=O)C1=CN(N=N1)O

Spécifications

Melting Point 105°C
Color Blanc
Quantity 1 g
Formula Weight 157.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material Ethyl 1-Hydroxy-1H-1,2,3-triazole-4-carboxylate