missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate 98.0+%, TCI America™
$104.38 - $341.13
Chemical Identifiers
| CAS | 100501-62-0 |
|---|---|
| Molecular Formula | C14H12F3NO3 |
| Molecular Weight (g/mol) | 299.25 |
| MDL Number | MFCD00276022 |
| InChI Key | LWLLHOVWIFISMG-UHFFFAOYSA-N |
| Synonym | ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid ethyl ester, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxohydroquinoline-3-carboxylic acid ethyl ester, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate dry wt. , may cont. up to water, acmc-1brcg, ksc499a1t, ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylate, neopentyl glycol-tetrahydrofuran copolymer, 2,2-dimethyl-1,3-propanediol-tetramethylene oxide copolymer |
| PubChem CID | 5140913 |
| IUPAC Name | ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate |
| SMILES | CCOC(=O)C1=CN(CC)C2=C(F)C(F)=C(F)C=C2C1=O |
Chemical Identifiers
| 100501-62-0 | |
| 299.25 | |
| LWLLHOVWIFISMG-UHFFFAOYSA-N | |
| 5140913 | |
| CCOC(=O)C1=CN(CC)C2=C(F)C(F)=C(F)C=C2C1=O |
| C14H12F3NO3 | |
| MFCD00276022 | |
| ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid ethyl ester, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxohydroquinoline-3-carboxylic acid ethyl ester, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate dry wt. , may cont. up to water, acmc-1brcg, ksc499a1t, ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylate, neopentyl glycol-tetrahydrofuran copolymer, 2,2-dimethyl-1,3-propanediol-tetramethylene oxide copolymer | |
| ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate |
Specifications
| 100501-62-0 | |
| White-Yellow | |
| MFCD00276022 | |
| ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid ethyl ester, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxohydroquinoline-3-carboxylic acid ethyl ester, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate dry wt. , may cont. up to water, acmc-1brcg, ksc499a1t, ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylate, neopentyl glycol-tetrahydrofuran copolymer, 2,2-dimethyl-1,3-propanediol-tetramethylene oxide copolymer | |
| CCOC(=O)C1=CN(CC)C2=C(F)C(F)=C(F)C=C2C1=O | |
| 299.25 | |
| 299.25 | |
| Crystalline Powder |
| 207°C | |
| C14H12F3NO3 | |
| 5 g | |
| LWLLHOVWIFISMG-UHFFFAOYSA-N | |
| ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate | |
| 5140913 | |
| ≥98.0% (GC) | |
| Ethyl 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate |
Safety and Handling
TSCA : No