missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate 98.0+%, TCI America™

Supplier:  TCI America E088825G

 View more versions of this product

Catalog No. E088825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate
207°C
C14H12F3NO3
25 g
LWLLHOVWIFISMG-UHFFFAOYSA-N
ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
5140913
≥98.0% (GC)
100501-62-0
White-Yellow
MFCD00276022
ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid ethyl ester, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxohydroquinoline-3-carboxylic acid ethyl ester, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate dry wt. , may cont. up to water, acmc-1brcg, ksc499a1t, ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylate, neopentyl glycol-tetrahydrofuran copolymer, 2,2-dimethyl-1,3-propanediol-tetramethylene oxide copolymer
CCOC(=O)C1=CN(CC)C2=C(F)C(F)=C(F)C=C2C1=O
299.25
299.25
Crystalline Powder

Chemical Identifiers

100501-62-0
299.25
LWLLHOVWIFISMG-UHFFFAOYSA-N
5140913
CCOC(=O)C1=CN(CC)C2=C(F)C(F)=C(F)C=C2C1=O
C14H12F3NO3
MFCD00276022
ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid ethyl ester, 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxohydroquinoline-3-carboxylic acid ethyl ester, ethyl 1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylate dry wt. , may cont. up to water, acmc-1brcg, ksc499a1t, ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylate, neopentyl glycol-tetrahydrofuran copolymer, 2,2-dimethyl-1,3-propanediol-tetramethylene oxide copolymer
ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions