missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Boc-piperazine-3-carboxylate, 97%

Catalog No. AAH6314603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6314603 1 g
AAH6314606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6314603 Supplier Thermo Scientific Chemicals Supplier No. H6314603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 183742-29-2 | C12H22N2O4 | 258.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 183742-29-2
Molecular Formula C12H22N2O4
Molecular Weight (g/mol) 258.318
MDL Number MFCD13172266
InChI Key IQKOVLZJPVVLOZ-UHFFFAOYSA-N
Synonym 1-tert-butyl 3-ethyl piperazine-1,3-dicarboxylate, 1-boc-3-piperazinecarboxylic acid ethyl ester, 1-n-boc-3-piperazinecarboxylic acid ethyl ester, 1-tert-butoxycarbonyl-3-ethoxycarbonylpiperazine, 1-tert-butyl3-ethylpiperazine-1,3-dicarboxylate, 1-tert-butyl-3-ethylpiperazine-1,3-dicarboxylate, ethyl 4-tert-butoxycarbonyl-2-piperazinecarboxylate, 1-1,1-dimethylethyl 3-ethyl 1,3-piperazinedicarboxylate, 1-tert-butoxycarbonylpiperazine-3-carboxylic acid ethyl ester, ethyl 4-1,1-dimethylethoxycarbonyl-2-piperazinecarboxylate
PubChem CID 11402671
IUPAC Name 1-O-tert-butyl 3-O-ethyl piperazine-1,3-dicarboxylate
SMILES CCOC(=O)C1CN(CCN1)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Formula Weight 258.31
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 1-Boc-piperazine-3-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only