accessibility menu, dialog, popup

UserName

H3COONHNOOCH3H3CCH3

CAS RN 183742-29-2

IUPAC Name: 1-tert-butyl 3-ethyl piperazine-1,3-dicarboxylate
Synonyms: 1-tert-butyl 3-ethyl piperazine-1,3-dicarboxylate
Molecular Weight (g/mol): 258.32
Molecular Formula: C12H22N2O4
InChi Key: IQKOVLZJPVVLOZ-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CN(CCN1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results