missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Boc-3-oxopiperidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH6410206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6410206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6410206 Supplier Thermo Scientific Chemicals Supplier No. H6410206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71233-25-5 | Molecular Formula: C13H21NO5 | Molecular Weight: 271.313 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71233-25-5
Molecular Formula C13H21NO5
Molecular Weight (g/mol) 271.313
MDL Number MFCD09878815
InChI Key WCTXJAXKORIYNA-UHFFFAOYSA-N
Synonym 1-tert-butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate, ethyl 1-n-boc-3-oxopiperidine-4-carboxylate, ethyl n-boc-3-oxopiperidine-4-carboxylate, 3-oxo-piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester, ethyl 1-n-boc-3-oxo-piperidine-4-carboxylate, ethyl 1-boc-3-oxopiperidine-4-carboxylate, ethyl n-tert-butoxycarbonyl-3-oxo-piperidine-4-carboxylate, 1-tert-butyl 4-ethyl 3-oxo-1,4-piperidinedicarboxylate, 1,4-piperidinedicarboxylic acid, 3-oxo-, 1-1,1-dimethylethyl 4-ethyl ester, ksc915i7p
PubChem CID 15852989
IUPAC Name 1-O-tert-butyl 4-O-ethyl 3-oxopiperidine-1,4-dicarboxylate
SMILES CCOC(=O)C1CCN(CC1=O)C(=O)OC(C)(C)C

Specifications

Quantity 5 g
Beilstein 421506
Formula Weight 271.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 1-Boc-3-oxopiperidine-4-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only