accessibility menu, dialog, popup

UserName

CAS RN 71233-25-5

IUPAC Name: 1-tert-butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate
Synonyms: 1-tert-butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate
Molecular Weight (g/mol): 271.31
Molecular Formula: C13H21NO5
InChi Key: WCTXJAXKORIYNA-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCN(CC1=O)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results