missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-benzylpyrrolidine-3-carboxylate, 97%

CAS: 5747-92-2 | C14H19NO2 | 233.311 g/mol

$317.75 - $1,286.90

Chemical Identifiers

CAS 5747-92-2
Molecular Formula C14H19NO2
Molecular Weight (g/mol) 233.311
MDL Number MFCD04039852
InChI Key CYPXEPWPTXKUPL-UHFFFAOYSA-N
Synonym 1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 1-benzyl-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, ethyl1-benzylpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid ethyl ester, acmc-1asr8, 1-benzyl-pyrrolidine-3-carboxylicacidethylester, ksc495s0t, 1-benzyl-3-carboethoxypyrrolidine, ethyl 1-benzyl pyrrolidine-3-carboxylate
PubChem CID 11622765
IUPAC Name ethyl 1-benzylpyrrolidine-3-carboxylate
SMILES CCOC(=O)C1CCN(C1)CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6496206
View Documents
Thermo Scientific Chemicals
H6496206
5 g N/A
Only null left
 
AAH6496214
View Documents
Thermo Scientific Chemicals
H6496214
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5747-92-2
233.311
CYPXEPWPTXKUPL-UHFFFAOYSA-N
11622765
CCOC(=O)C1CCN(C1)CC2=CC=CC=C2
C14H19NO2
MFCD04039852
1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 1-benzyl-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, ethyl1-benzylpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid ethyl ester, acmc-1asr8, 1-benzyl-pyrrolidine-3-carboxylicacidethylester, ksc495s0t, 1-benzyl-3-carboethoxypyrrolidine, ethyl 1-benzyl pyrrolidine-3-carboxylate
ethyl 1-benzylpyrrolidine-3-carboxylate

Specifications

5747-92-2
C14H19NO2
5 g
1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 1-benzyl-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, ethyl1-benzylpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid ethyl ester, acmc-1asr8, 1-benzyl-pyrrolidine-3-carboxylicacidethylester, ksc495s0t, 1-benzyl-3-carboethoxypyrrolidine, ethyl 1-benzyl pyrrolidine-3-carboxylate
CCOC(=O)C1CCN(C1)CC2=CC=CC=C2
233.311
233.31
Liquid
Yellow
MFCD04039852
398388
CYPXEPWPTXKUPL-UHFFFAOYSA-N
ethyl 1-benzylpyrrolidine-3-carboxylate
11622765
97%
Ethyl 1-benzylpyrrolidine-3-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only