CAS RN 5747-92-2
IUPAC Name:
ethyl 1-benzylpyrrolidine-3-carboxylate
Synonyms:
ethyl 1-benzylpyrrolidine-3-carboxylate
Molecular Weight (g/mol):
233.31
Molecular Formula:
C14H19NO2
InChi Key:
CYPXEPWPTXKUPL-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1CCN(CC2=CC=CC=C2)C1
1
–
1
of
1
results
1
Ethyl 1-benzylpyrrolidine-3-carboxylate, 97%
CAS: 5747-92-2 Molecular Formula: C14H19NO2 Molecular Weight (g/mol): 233.311 MDL Number: MFCD04039852 InChI Key: CYPXEPWPTXKUPL-UHFFFAOYSA-N Synonym: 1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester,ethyl 1-benzyl-pyrrolidine-3-carboxylate,3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester,ethyl1-benzylpyrrolidine-3-carboxylate,1-benzylpyrrolidine-3-carboxylic acid ethyl ester,acmc-1asr8,1-benzyl-pyrrolidine-3-carboxylicacidethylester,ksc495s0t,1-benzyl-3-carboethoxypyrrolidine,ethyl 1-benzyl pyrrolidine-3-carboxylate PubChem CID: 11622765 IUPAC Name: ethyl 1-benzylpyrrolidine-3-carboxylate SMILES: CCOC(=O)C1CCN(C1)CC2=CC=CC=C2