missing translation for 'onlineSavingsMsg'
Learn More

Éthyle 1-Adamantanecarboxylate 98,0+%, TCI America™

Numéro de catalogue. A07225G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A07225G 5 g
A072225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A07225G Fournisseur TCI America Code fournisseur A07225G

Identifiants chimiques

CAS 2094-73-7
Molecular Formula C13H20O2
Molecular Weight (g/mol) 208.30
MDL Number MFCD00078272
InChI Key SYEXGNJRYPOUSI-UHFFFAOYSA-N
Synonym 1-adamantanecarboxylic acid ethyl ester, ethyl 1-adamantanecarboxylate, ethyladamantane-1-carboxylate, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, ethyl ester, ethyl tricyclo 3.3.1.13,7 decane-1-carboxylate, tricyclo 3.3.1.1 3,7-decane-1-carboxylic acid, ethyl ester, pubchem8778, acmc-1cctj, 1-ethoxycarbonyladamantane, ethyl adamantanecarboxylate
PubChem CID 123141
IUPAC Name Éthyle adamantane-1-carboxylate
SMILES CCOC(=O)C12CC3CC(CC(C3)C1)C2

Spécifications

Color Incolore
Boiling Point 123°C
Quantity 5 g
Formula Weight 208.30
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material Ethyl 1-Adamantanecarboxylate