CAS RN 2094-73-7
IUPAC Name:
ethyl adamantane-1-carboxylate
Synonyms:
ethyl tricyclodecanecarboxylate
Molecular Weight (g/mol):
208.3
Molecular Formula:
C13H20O2
InChi Key:
SYEXGNJRYPOUSI-UHFFFAOYSA-N
SMILES:
CCOC(=O)C12CC3CC(CC(C3)C1)C2
1
–
1
of
1
results
1
Ethyl 1-Adamantanecarboxylate 98.0+%, TCI America™
CAS: 2094-73-7 Molecular Formula: C13H20O2 Molecular Weight (g/mol): 208.30 MDL Number: MFCD00078272 InChI Key: SYEXGNJRYPOUSI-UHFFFAOYSA-N Synonym: 1-adamantanecarboxylic acid ethyl ester,ethyl 1-adamantanecarboxylate,ethyladamantane-1-carboxylate,tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, ethyl ester,ethyl tricyclo 3.3.1.13,7 decane-1-carboxylate,tricyclo 3.3.1.1 3,7-decane-1-carboxylic acid, ethyl ester,pubchem8778,acmc-1cctj,1-ethoxycarbonyladamantane,ethyl adamantanecarboxylate PubChem CID: 123141 IUPAC Name: ethyl adamantane-1-carboxylate SMILES: CCOC(=O)C12CC3CC(CC(C3)C1)C2