missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1,4-Benzodioxane-2-carboxylate 98.0+%, TCI America™
Supplier: TCI America E090225G
Specifications
| Ethyl 1,4-Benzodioxane-2-carboxylate | |
| Yellow | |
| C11H12O4 | |
| 25 g | |
| DDIGEMWIKJMEIU-UHFFFAOYSA-N | |
| ethyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate | |
| 151226 | |
| ≥98.0% (GC) |
| 4739-94-0 | |
| 115°C | |
| MFCD01099774 | |
| 1,4-Benzodioxane-2-carboxylic Acid Ethyl Ester | |
| CCOC(=O)C1COC2=CC=CC=C2O1 | |
| 208.213 | |
| 208.21 | |
| Liquid |
Chemical Identifiers
| 4739-94-0 | |
| 208.213 | |
| DDIGEMWIKJMEIU-UHFFFAOYSA-N | |
| 151226 | |
| CCOC(=O)C1COC2=CC=CC=C2O1 |
| C11H12O4 | |
| MFCD01099774 | |
| 1,4-Benzodioxane-2-carboxylic Acid Ethyl Ester | |
| ethyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate |
Safety and Handling
TSCA : No