accessibility menu, dialog, popup

UserName

CH3OOOO

CAS RN 4739-94-0

IUPAC Name: ethyl 2,3-dihydro-1,4-benzodioxine-2-carboxylate
Synonyms: ethyl 2,3-dihydro-1,4-benzodioxine-2-carboxylate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: DDIGEMWIKJMEIU-UHFFFAOYNA-N
SMILES: CCOC(=O)C1COC2=CC=CC=C2O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results