missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,4-Benzodioxane-2-carboxylate 98.0+%, TCI America™

Catalog No. E09025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09025G 5 g
E090225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09025G Supplier TCI America Supplier No. E09025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4739-94-0
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD01099774
InChI Key DDIGEMWIKJMEIU-UHFFFAOYSA-N
Synonym 1,4-Benzodioxane-2-carboxylic Acid Ethyl Ester
PubChem CID 151226
IUPAC Name ethyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate
SMILES CCOC(=O)C1COC2=CC=CC=C2O1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 208.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1,4-Benzodioxane-2-carboxylate