missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,1,2,2-Tetrafluoroethyl Ether 98.0+%, TCI America™

Supplier:  TCI America E101925G

 View more versions of this product

Catalog No. E101925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 1,1,2,2-Tetrafluoroethyl Ether
Colorless
C4H6F4O
25 g
ethyl 1,1,2,2-tetrafluoroethyl ether, 374pce-beta-gamma, 1,1,2,2-tetrafluoroethyl ethyl ether, ethane, 1-ethoxy-1,1,2,2-tetrafluoro, ether, ethyl 1,1,2,2-tetrafluoroethyl, acmc-1anqb, 1,1,2,2-tetrafluoroethyl ether, 1,1,2,2-tetrafluoroethoxyethane, 1,1,2,2-tetrafluoro ethyl ether, 3-oxa-1,1,2,2-tetrafluoropentane
CCOC(F)(F)C(F)F
146.09
146.08
Liquid
512-51-6
58°C
MFCD00069162
3271
HBRLMDFVVMYNFH-UHFFFAOYSA-N
1-ethoxy-1,1,2,2-tetrafluoroethane
120272
≥98.0% (GC)

Chemical Identifiers

512-51-6
146.09
HBRLMDFVVMYNFH-UHFFFAOYSA-N
120272
CCOC(F)(F)C(F)F
C4H6F4O
MFCD00069162
ethyl 1,1,2,2-tetrafluoroethyl ether, 374pce-beta-gamma, 1,1,2,2-tetrafluoroethyl ethyl ether, ethane, 1-ethoxy-1,1,2,2-tetrafluoro, ether, ethyl 1,1,2,2-tetrafluoroethyl, acmc-1anqb, 1,1,2,2-tetrafluoroethyl ether, 1,1,2,2-tetrafluoroethoxyethane, 1,1,2,2-tetrafluoro ethyl ether, 3-oxa-1,1,2,2-tetrafluoropentane
1-ethoxy-1,1,2,2-tetrafluoroethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : KI3590000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions