accessibility menu, dialog, popup

UserName

CH3OFFFF

CAS RN 512-51-6

IUPAC Name: 1-ethoxy-1,1,2,2-tetrafluoroethane
Synonyms: 1-ethoxy-1,1,2,2-tetrafluoroethane
Molecular Weight (g/mol): 146.09
Molecular Formula: C4H6F4O
InChi Key: HBRLMDFVVMYNFH-UHFFFAOYSA-N
SMILES: CCOC(F)(F)C(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results