missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,1,2,2-Tetrafluoroethyl Ether 98.0+%, TCI America™

Catalog No. E10195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10195G 5 g
E101925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10195G Supplier TCI America Supplier No. E10195G

Chemical Identifiers

CAS 512-51-6
Molecular Formula C4H6F4O
Molecular Weight (g/mol) 146.09
MDL Number MFCD00069162
InChI Key HBRLMDFVVMYNFH-UHFFFAOYSA-N
Synonym ethyl 1,1,2,2-tetrafluoroethyl ether, 374pce-beta-gamma, 1,1,2,2-tetrafluoroethyl ethyl ether, ethane, 1-ethoxy-1,1,2,2-tetrafluoro, ether, ethyl 1,1,2,2-tetrafluoroethyl, acmc-1anqb, 1,1,2,2-tetrafluoroethyl ether, 1,1,2,2-tetrafluoroethoxyethane, 1,1,2,2-tetrafluoro ethyl ether, 3-oxa-1,1,2,2-tetrafluoropentane
PubChem CID 120272
IUPAC Name 1-ethoxy-1,1,2,2-tetrafluoroethane
SMILES CCOC(F)(F)C(F)F

Specifications

Color Colorless
Boiling Point 58°C
Quantity 5 g
UN Number 3271
Formula Weight 146.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1,1,2,2-Tetrafluoroethyl Ether
RTECSNumber KI3590000
TSCA No