missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethoxycarbonyl Isothiocyanate 95.0+%, TCI America™
Supplier: TCI America E044025G
Specifications
| Ethoxycarbonyl Isothiocyanate | |
| 61°C | |
| MFCD00004814 | |
| 1992 | |
| BDTDECDAHYOJRO-UHFFFAOYSA-N | |
| ethyl N-(sulfanylidenemethylidene)carbamate | |
| 85320 | |
| ≥95.0% (GC) |
| 16182-04-0 | |
| C4H5NO2S | |
| 25 g | |
| ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate | |
| CCOC(=O)N=C=S | |
| 131.149 | |
| 131.15 | |
| Liquid |
Chemical Identifiers
| 16182-04-0 | |
| 131.149 | |
| BDTDECDAHYOJRO-UHFFFAOYSA-N | |
| 85320 | |
| CCOC(=O)N=C=S |
| C4H5NO2S | |
| MFCD00004814 | |
| ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate | |
| ethyl N-(sulfanylidenemethylidene)carbamate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator