accessibility menu, dialog, popup

UserName

CH3OONCS

CAS RN 16182-04-0

IUPAC Name: ethyl N-carbothioylcarbamate
Synonyms: ethyl N-carbothioylcarbamate
Molecular Weight (g/mol): 131.15
Molecular Formula: C4H5NO2S
InChi Key: BDTDECDAHYOJRO-UHFFFAOYSA-N
SMILES: CCOC(=O)N=C=S
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)