missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonyl Isothiocyanate 95.0+%, TCI America™

Catalog No. E04405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04405G 5 g
E044025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04405G Supplier TCI America Supplier No. E04405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16182-04-0
Molecular Formula C4H5NO2S
Molecular Weight (g/mol) 131.149
MDL Number MFCD00004814
InChI Key BDTDECDAHYOJRO-UHFFFAOYSA-N
Synonym ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate
PubChem CID 85320
IUPAC Name ethyl N-(sulfanylidenemethylidene)carbamate
SMILES CCOC(=O)N=C=S

Specifications

Boiling Point 61°C
Quantity 5 g
UN Number 1992
Formula Weight 131.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethoxycarbonyl Isothiocyanate
TSCA No
Recommended Storage Refrigerator