Learn More
Ethoxycarbonyl isothiocyanate, 95%, Thermo Scientific™
$126.19 - $469.60
Chemical Identifiers
| CAS | 16182-04-0 |
|---|---|
| Molecular Formula | C4H5NO2S |
| Molecular Weight (g/mol) | 131.15 |
| MDL Number | MFCD00004814 |
| InChI Key | BDTDECDAHYOJRO-UHFFFAOYSA-N |
| Synonym | ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate |
| PubChem CID | 85320 |
| IUPAC Name | ethyl N-(sulfanylidenemethylidene)carbamate |
| SMILES | CCOC(=O)N=C=S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC187850050
|
N/A | 5g | Glass bottle |
Each for $126.19
|
|
||||
|
AC187850250
|
N/A | 25g | Glass bottle |
Each for $469.60
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 16182-04-0 | |
| 131.15 | |
| BDTDECDAHYOJRO-UHFFFAOYSA-N | |
| 85320 | |
| CCOC(=O)N=C=S |
| C4H5NO2S | |
| MFCD00004814 | |
| ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate | |
| ethyl N-(sulfanylidenemethylidene)carbamate |
Specifications
| 16182-04-0 | |
| 56.0°C (18.0 mmHg) | |
| 94% min. (GC) | |
| SCNCOOCH2CH3 | |
| 03, III, 279 | |
| 1.16 | |
| BDTDECDAHYOJRO-UHFFFAOYSA-N | |
| ethyl N-(sulfanylidenemethylidene)carbamate | |
| 85320 | |
| 95% |
| 1.1600g/mL | |
| 50°C | |
| C4H5NO2S | |
| MFCD00004814 | |
| 04,223; 06,250; 07,146 | |
| ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate | |
| CCOC(=O)N=C=S | |
| 131.15 | |
| 131.15 | |
| Ethoxycarbonyl isothiocyanate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Danger
EINECSNumber : 240-318-9
RUO – Research Use Only