missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyacetic Acid 98.0+%, TCI America™

Supplier:  TCI America E0377100G

 View more versions of this product

Catalog No. E0377100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethoxyacetic Acid
Yellow
C4H8O3
100 g
ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
CCOCC(=O)O
104.105
104.11
Liquid
627-03-2
207°C
MFCD00004310
3265
YZGQDNOIGFBYKF-UHFFFAOYSA-N
2-ethoxyacetic acid
12301
≥98.0% (GC,T)

Chemical Identifiers

627-03-2
104.105
YZGQDNOIGFBYKF-UHFFFAOYSA-N
12301
CCOCC(=O)O
C4H8O3
MFCD00004310
ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
2-ethoxyacetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3131

RTECSNumber : AH3600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions