missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethoxyacetic Acid 98.0+%, TCI America™
$96.94 - $779.17
Chemical Identifiers
| CAS | 627-03-2 |
|---|---|
| Molecular Formula | C4H8O3 |
| Molecular Weight (g/mol) | 104.105 |
| MDL Number | MFCD00004310 |
| InChI Key | YZGQDNOIGFBYKF-UHFFFAOYSA-N |
| Synonym | ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 |
| PubChem CID | 12301 |
| IUPAC Name | 2-ethoxyacetic acid |
| SMILES | CCOCC(=O)O |
Chemical Identifiers
| 627-03-2 | |
| 104.105 | |
| YZGQDNOIGFBYKF-UHFFFAOYSA-N | |
| 12301 | |
| CCOCC(=O)O |
| C4H8O3 | |
| MFCD00004310 | |
| ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 | |
| 2-ethoxyacetic acid |
Specifications
| 627-03-2 | |
| 207°C | |
| MFCD00004310 | |
| 3265 | |
| YZGQDNOIGFBYKF-UHFFFAOYSA-N | |
| 2-ethoxyacetic acid | |
| 12301 | |
| ≥98.0% (GC,T) | |
| Ethoxyacetic Acid |
| Yellow | |
| C4H8O3 | |
| 25 g | |
| ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294 | |
| CCOCC(=O)O | |
| 104.105 | |
| 104.11 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3131
RTECSNumber : AH3600000
TSCA : Yes