missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethisterone 98.0+%, TCI America™
$44.73 - $117.43
Chemical Identifiers
| CAS | 434-03-7 |
|---|---|
| Molecular Formula | C21H28O2 |
| Molecular Weight (g/mol) | 312.453 |
| MDL Number | MFCD00003656 |
| InChI Key | CHNXZKVNWQUJIB-SJFWLOONSA-N |
| Synonym | pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one |
| PubChem CID | 12016247 |
| IUPAC Name | (8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one |
| SMILES | CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C |
Chemical Identifiers
| 434-03-7 | |
| 312.453 | |
| CHNXZKVNWQUJIB-SJFWLOONSA-N | |
| 12016247 | |
| CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C |
| C21H28O2 | |
| MFCD00003656 | |
| pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one | |
| (8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one |
Specifications
| 434-03-7 | |
| C21H28O2 | |
| 1 g | |
| CHNXZKVNWQUJIB-SJFWLOONSA-N | |
| (8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one | |
| 12016247 | |
| ≥98.0% (HPLC,T) | |
| Ethisterone |
| 272°C | |
| MFCD00003656 | |
| pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one | |
| CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C | |
| 312.453 | |
| 312.45 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TU5570250
TSCA : No