missing translation for 'onlineSavingsMsg'
Learn More

Ethinyl Estradiol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017812
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017812 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017812 Supplier Merck Emd Millipore Supplier No. PHR1480200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 57-63-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H24O2
MDL Number MFCD00003690
Melting Point 182°C to 183°C
Synonym 17 alpha-Ethynylestradiol; 17 alpha-Ethynyl-1,3,5(10)-estratriene-3,17 beta-diol; 19-Nor-1,3,5(10),17 alpha-pregnatrien-20-yne-3,17-diol; Ethinylestradiol
Type Certified Reference Material
Quantity 200 mg
Formula Weight 296.40
Grade Certified Reference Material