missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide soln., 10mg/ml

CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol

Supplier:  Thermo Scientific Chemicals J62282AB

Catalog No. AAJ62282AB


Only null left

Description

Description

Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.1,2,3 It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA.4 Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase. Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethidium bromide soln.
1239-45-8
MFCD00011724
10mg/ml
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
ZMMJGEGLRURXTF-UHFFFAOYSA-N
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
14710
394.32
10 mL
Red
C21H20BrN3
UN2810
14,4731
Soluble in water (10mg/mL).
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
394.32
CHEBI:4883
Odorless
Liquid

Chemical Identifiers

1239-45-8
394.32
ZMMJGEGLRURXTF-UHFFFAOYSA-N
14710
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
C21H20BrN3
MFCD00011724
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
CHEBI:4883
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P261-P271-P281-P304+P340-P308+P313-P311-P501c

H331-H341

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 214-984-6

RTECSNumber : SF7950000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only