missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide, 95%, pure

CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol

Supplier:  Thermo Scientific Chemicals 170960050

 View more versions of this product

Catalog No. AC170960050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethidium bromide
260.0°C to 262.0°C
∼4.5 (1% aq. soln. at 20°C)
5.0% max.
94% min. (Argentometry)
C21H20BrN3
5 g
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
ZMMJGEGLRURXTF-UHFFFAOYSA-N
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
14710
394.3
Pure
1239-45-8
Purple to Red
>100°C
Authentic
Glass bottle
MFCD00011724
15,4769
(1% in water) Clear red
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
394.32
CHEBI:4883
95%
Crystalline Powder

Chemical Identifiers

1239-45-8
394.32
ZMMJGEGLRURXTF-UHFFFAOYSA-N
14710
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
C21H20BrN3
MFCD00011724
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
CHEBI:4883
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Fatal if inhaled.
Harmful if swallowed.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 214-984-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only