missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine Thioglycolate (ca. 85% in Water), TCI America™

Catalog No. E081325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E081325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E081325G Supplier TCI America Supplier No. E081325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 126-97-6
Molecular Formula C4H11NO3S
Molecular Weight (g/mol) 153.196
MDL Number MFCD00050789
InChI Key XTJCJAPNPGGFED-UHFFFAOYSA-N
Synonym Monoethanolamine Thioglycolate, Ethanolamine Mercaptoacetate
PubChem CID 160430
IUPAC Name 2-aminoethanol;2-sulfanylacetic acid
SMILES C(CO)N.C(C(=O)O)S

Specifications

Color Colorless
Quantity 25 g
Formula Weight 153.20
Physical Form Liquid
Chemical Name or Material Ethanolamine Thioglycolate (ca. 85% in Water)